Please login to your online account to display your discounted pricing

Lanthanum(III) nitrate hexahydrate, 98+%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  218715000

 View more versions of this product

Catalog No. AC218715000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.0% (Complexometry)
Formula Weight 433.01
Physical Form Powder
Quantity 500g
Color White to light yellow
RTECS Number OE5250000
Fieser 11,283
Merck Index 15, 5414
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May intensify fire; oxidizer.
Health Hazard 1 Danger
CAS 10277-43-7
MDL Number MFCD00149751
Chemical Name or Material Lanthanum(III) nitrate hexahydrate
Assay Percent Range 98+%
Name Note 98+%
Linear Formula La(NO3)3·6H2O
Molecular Formula LaN3O9·6H2O
Other Impurities Ce02: < 0.1%, Pr6O11: < 0.1%, Nd2O3: < 0.1%
Fe2O3 < 0.001%
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Lanthanum(III) nitrate hexahydrate
Name Note 98+%
CAS 10277-43-7
Assay ≥98.0% (Complexometry)
Assay Percent Range 98+%
Molecular Formula LaN3O9·6H2O
Linear Formula La(NO3)3·6H2O
Formula Weight 433.01
MDL Number MFCD00149751
Accessories

Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May intensify fire; oxidizer.

SDS & Safety Documents