missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lanthanum(III) Isopropoxide, TCI America™

Manufacturer:  TCI America L0174200MG

 View more versions of this product

Catalog No. L0174200MG



Specifications

Specifications

Lanthanum(III) Isopropoxide
White-Yellow
200mg
C9H21LaO3
1325
SORGMJIXNUWMMR-UHFFFAOYSA-N
lanthanum(3+);propan-2-olate
9858075
Crystal-Powder at 20°C
300°C
128°C
19446-52-7
MFCD00015305
lanthanum isopropoxide, lanthanum iii isopropoxide, acmc-20ako6, lanthanum triisopropoxide, lanthanum 3+ ion tris propan-2-olate, lanthanum 3+ tris propan-2-olate
CC(C)[O-].CC(C)[O-].CC(C)[O-].[La+3]
316.169
316.17

Chemical Identifiers

19446-52-7
316.169
SORGMJIXNUWMMR-UHFFFAOYSA-N
9858075
CC(C)[O-].CC(C)[O-].CC(C)[O-].[La+3]
C9H21LaO3
MFCD00015305
lanthanum isopropoxide, lanthanum iii isopropoxide, acmc-20ako6, lanthanum triisopropoxide, lanthanum 3+ ion tris propan-2-olate, lanthanum 3+ tris propan-2-olate
lanthanum(3+);propan-2-olate
Safety and Handling

Safety and Handling

TSCA : No

Documents