missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lanthanum(III) Isopropoxide, TCI America™

$62.55 - $217.25

Chemical Identifiers

CAS 19446-52-7
Molecular Formula C9H21LaO3
Molecular Weight (g/mol) 316.169
MDL Number MFCD00015305
InChI Key SORGMJIXNUWMMR-UHFFFAOYSA-N
Synonym lanthanum isopropoxide, lanthanum iii isopropoxide, acmc-20ako6, lanthanum triisopropoxide, lanthanum 3+ ion tris propan-2-olate, lanthanum 3+ tris propan-2-olate
PubChem CID 9858075
IUPAC Name lanthanum(3+);propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].[La+3]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
L01741G View Documents Tci America
L01741G
1g Each for $217.25
 
L0174200MG View Documents Tci America
L0174200MG
200mg Each for $62.55
 
Specifications

Chemical Identifiers

19446-52-7
316.169
SORGMJIXNUWMMR-UHFFFAOYSA-N
9858075
CC(C)[O-].CC(C)[O-].CC(C)[O-].[La+3]
C9H21LaO3
MFCD00015305
lanthanum isopropoxide, lanthanum iii isopropoxide, acmc-20ako6, lanthanum triisopropoxide, lanthanum 3+ ion tris propan-2-olate, lanthanum 3+ tris propan-2-olate
lanthanum(3+);propan-2-olate

Specifications

300°C
128°C
19446-52-7
MFCD00015305
lanthanum isopropoxide, lanthanum iii isopropoxide, acmc-20ako6, lanthanum triisopropoxide, lanthanum 3+ ion tris propan-2-olate, lanthanum 3+ tris propan-2-olate
CC(C)[O-].CC(C)[O-].CC(C)[O-].[La+3]
316.169
316.17
Lanthanum(III) Isopropoxide
White-Yellow
1g
C9H21LaO3
1325
SORGMJIXNUWMMR-UHFFFAOYSA-N
lanthanum(3+);propan-2-olate
9858075
Crystal-Powder at 20°C
Safety and Handling

Safety and Handling

TSCA : No