Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum(III) isopropoxide, La 40%

Chemical Identifiers

Chemical Name or Material Lanthanum(III) isopropoxide
CAS 19446-52-7
Linear Formula La[OCH(CH3)2]3
Formula Weight 316.18
MDL Number MFCD00015305
UN Number UN2925
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1461602

  SDS
alfa aesar™
1461602
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1461604

  SDS
alfa aesar™
1461604
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1461609

  SDS
alfa aesar™
1461609
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 316.18
Health Hazard 2 GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Health Hazard 1 Danger
Physical Form Powder
Quantity 0.5g
Odor Alcohol-like
CAS 19446-52-7
Boiling Point 170°C (0.04mmHg) (subl.)
MDL Number MFCD00015305
Linear Formula La[OCH(CH3)2]3
Melting Point 120° to 128°C
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
UN Number UN2925
Sensitivity Air & Moisture Sensitive
Chemical Name or Material Lanthanum(III) isopropoxide
Health Hazard 3 GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Color White
Concentration La 40%
Safety and Handling

Danger

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.