missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum(III) isopropoxide, La 40%

Manufacturer:  Alfa Aesar™ 1461609

 View more versions of this product

Catalog No. AA1461609



Specifications

Specifications

Lanthanum(III) isopropoxide
Air & Moisture Sensitive
White
10g
C9H21LaO3
MFCD00015305
lanthanum isopropoxide, lanthanum iii isopropoxide, acmc-20ako6, lanthanum triisopropoxide, lanthanum 3+ ion tris propan-2-olate, lanthanum 3+ tris propan-2-olate
SORGMJIXNUWMMR-UHFFFAOYSA-N
lanthanum(3+);propan-2-olate
9858075
Powder
Alcohol-like
170°C (0.04mmHg)(sublimation)
120°C to 128°C
19446-52-7
La[OCH(CH3)2]3
UN2925
La 40%
CC(C)[O-].CC(C)[O-].CC(C)[O-].[La+3]
316.169
316.18

Chemical Identifiers

19446-52-7
316.169
SORGMJIXNUWMMR-UHFFFAOYSA-N
9858075
CC(C)[O-].CC(C)[O-].CC(C)[O-].[La+3]
C9H21LaO3
MFCD00015305
lanthanum isopropoxide, lanthanum iii isopropoxide, acmc-20ako6, lanthanum triisopropoxide, lanthanum 3+ ion tris propan-2-olate, lanthanum 3+ tris propan-2-olate
lanthanum(3+);propan-2-olate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.

SDS Documents