Please login to your online account to display your discounted pricing

Lanthanum(III) Isopropoxide, 98%, ACROS Organics™


Quantity 1g
Packaging Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  AC383640010

 View more versions of this product

Catalog No. AC383640010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 316.17
Quantity 1g
Color Off-white to pale yellow
Packaging Glass Bottle in Tin Can
Linear Formula [(CH3)2CHO)3La
Health Hazard 1 GHS Signal Word: Danger
Assay Percent Range 98%
Molecular Formula C9H21LaO3
CAS 19446-52-7
Identification Positive for isopropoxy moiety (GC)
Health Hazard 2 GHS H Statement:
Flammable solid.
In contact with water releases flammable gases.
Causes severe skin burns and eye damage.
May be corrosive to metals.
MDL Number MFCD00015305
Chemical Name or Material Lanthanum(III) isopropoxide
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Lanthanum(III) isopropoxide
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H21LaO3
Linear Formula [(CH3)2CHO)3La
Formula Weight 316.17
MDL Number MFCD00015305
Accessories

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable solid.
In contact with water releases flammable gases.
Causes severe skin burns and eye damage.
May be corrosive to metals.

SDS & Safety Documents