Please login to your online account to display your discounted pricing

Lanthanum(III) Isopropoxide, 98%, ACROS Organics™

$120.04 - $372.87

Chemical Identifiers

Chemical Name or Material Lanthanum(III) isopropoxide
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H21LaO3
Linear Formula [(CH3)2CHO)3La
Formula Weight 316.17
MDL Number MFCD00015305
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC383640010

  SDS
thermo fisher scientific
AC383640010
Glass Bottle in Tin Can 1g Each for $120.04

AC383640050

  SDS
thermo fisher scientific
AC383640050
Glass bottle 5g Each for $372.87
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 316.17
Quantity 1g
Color Off-white to pale yellow
Packaging Glass Bottle in Tin Can
Linear Formula [(CH3)2CHO)3La
Health Hazard 1 GHS Signal Word: Danger
Assay Percent Range 98%
Molecular Formula C9H21LaO3
CAS 19446-52-7
Identification Positive for isopropoxy moiety (GC)
Health Hazard 2 GHS H Statement:
Flammable solid.
In contact with water releases flammable gases.
Causes severe skin burns and eye damage.
May be corrosive to metals.
MDL Number MFCD00015305
Chemical Name or Material Lanthanum(III) isopropoxide
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable solid.
In contact with water releases flammable gases.
Causes severe skin burns and eye damage.
May be corrosive to metals.