Learn More
Lanthanum(III) Isopropoxide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 383640050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lanthanum(III) isopropoxide | |
White to Yellow | |
Glass bottle | |
[(CH3)2CHO]3La | |
5 g | |
Solubility in water: reacts | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].[La+3] | |
316.17 | |
316.17 | |
Powder |
19446-52-7 | |
97.5% min. (Complexometry) | |
C9H21LaO3 | |
MFCD00015305 | |
lanthanum isopropoxide, lanthanum iii isopropoxide, acmc-20ako6, lanthanum triisopropoxide, lanthanum 3+ ion tris propan-2-olate, lanthanum 3+ tris propan-2-olate | |
SORGMJIXNUWMMR-UHFFFAOYSA-N | |
lanthanum(3+);propan-2-olate | |
9858075 | |
98% |
Chemical Identifiers
19446-52-7 | |
316.17 | |
SORGMJIXNUWMMR-UHFFFAOYSA-N | |
9858075 | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].[La+3] |
C9H21LaO3 | |
MFCD00015305 | |
lanthanum isopropoxide, lanthanum iii isopropoxide, acmc-20ako6, lanthanum triisopropoxide, lanthanum 3+ ion tris propan-2-olate, lanthanum 3+ tris propan-2-olate | |
lanthanum(3+);propan-2-olate |
Safety and Handling
GHS H Statement:
Flammable solid.
In contact with water releases flammable gases.
Causes severe skin burns and eye damage.
May be corrosive to metals.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only