missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum(III) iodide, ultra dry, 99.995+% (metals basis)

Manufacturer:  Alfa Aesar™ 4454803

 View more versions of this product

Catalog No. AA4454803



Description

Description

  • Ampouled under argon
Specifications

Specifications

Lanthanum (III) iodide
13813-22-4
LaI3
519.62
≥99.995%
Ampule
772°C
Ultra dry
(metals basis)
MFCD00049470
-10 Mesh Beads,Ampouled under argon
−10 Mesh
Moisture sensitive
1g

Chemical Identifiers

13813-22-4
LaI3
MFCD00049470
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 237-465-6

TSCA : Yes

storageNote1 :

  • Ampouled under argon

Recommended Storage : Ambient temperatures

Documents