missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum(III) iodide, ultra dry, 99.995+% (metals basis)

Manufacturer:  Alfa Aesar™ 4454806

 View more versions of this product

Catalog No. AA4454806



Description

Description

  • Ampouled under argon
Specifications

Specifications

Lanthanum (III) iodide
13813-22-4
LaI3
519.62
≥99.995%
Ampule
772°C
Ultra dry
(metals basis)
MFCD00049470
Beads
−10 Mesh
Moisture Sensitive
5g

Chemical Identifiers

13813-22-4
LaI3
MFCD00049470
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

storageNote1 :

  • Ampouled under argon

SDS Documents