Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum(III) fluoride, Optical Grade

Chemical Identifiers

Chemical Name or Material Lanthanum(III) fluoride
CAS 13709-38-1
Purity Grade Optical Grade
Linear Formula LaF3
Formula Weight 195.9
MDL Number MFCD00011073
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1406103

  SDS
alfa aesar™
1406103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1406106

  SDS
alfa aesar™
1406106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 195.9
Linear Formula LaF3
Physical Form Broken Crystals
Purity Grade Optical Grade
Quantity 1g
Chemical Name or Material Lanthanum(III) fluoride
CAS 13709-38-1
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 5.936g/mL
Sensitivity Hygroscopic
MDL Number MFCD00011073
TSCA TSCA
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Odor Odorless
Melting Point 1493°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.