Please login to your online account to display your discounted pricing

Lanthanum(III) fluoride, anhydrous, 99.99% (REO), Alfa Aesar™, REacton™

Chemical Identifiers

Chemical Name or Material Lanthanum(III) fluoride, anhydrous
CAS 13709-38-1
Physical Form Crystals
Assay Percent Range 99.99% (REO)
Molecular Formula F3La
Formula Weight 195.9
MDL Number MFCD00011073
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1127318

  SDS
alfa aesar
1127318
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1127330

  SDS
alfa aesar
1127330
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystals
Formula Weight 195.9
Quantity 50g
Color White
EINECS Number 237-252-8
Assay Percent Range 99.99% (REO)
MDL Number MFCD00011073
Chemical Name or Material Lanthanum(III) fluoride, anhydrous
Health Hazard 1 H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Molecular Formula F3La
Appearance White crystalline
Odor Odorless
CAS 13709-38-1
Safety and Handling

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.