Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum(III) ethoxide, 95% (REO)

Chemical Identifiers

Chemical Name or Material Lanthanum(III) ethoxide
CAS 90397-54-9
Linear Formula La(OCH2CH3)3
Formula Weight 274.08
MDL Number MFCD00075070
UN Number UN2925
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3579104

 
alfa aesar™
3579104
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3579109

 
alfa aesar™
3579109
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 274.08
Chemical Name or Material Lanthanum(III) ethoxide
Physical Form Crystalline
Assay 95%
Assay Percent Notes (REO)
Linear Formula La(OCH2CH3)3
MDL Number MFCD00075070
CAS 90397-54-9
UN Number UN2925
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Health Hazard 3 GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Color White
Quantity 2g
Sensitivity Moisture Sensitive
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.