Please login to your online account to display your discounted pricing

Lanthanum(III) chloride, ultra dry, White beads, 99.99% (Metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Lanthanum(III) chloride, ultra dry
CAS 10099-58-8
Physical Form Beads
Assay Percent Range 99.99% (Metals basis)
Molecular Formula LaCl3
Formula Weight 245.27
MDL Number MFCD00011068
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4432206

  SDS
alfa aesar
4432206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4432214

  SDS
alfa aesar
4432214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Beads
Formula Weight 245.27
Quantity 5g
Color White
EINECS Number 233-237-5
Chemical Name or Material Lanthanum(III) chloride, ultra dry
Odor Odorless
Assay Percent Range 99.99% (Metals basis)
Molecular Formula LaCl3
MDL Number MFCD00011068
CAS 10099-58-8
Health Hazard 1 H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Safety and Handling

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.