missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum(III) chloride, ultra dry, 99.9% (REO)

Chemical Identifiers

CAS 10099-58-8
Formula Weight 245.27
MDL Number MFCD00011068
UN Number UN3077
Merck Index 14,5363
Chemical Name or Material Lanthanum(III) chloride, Ultra dry
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3570209 SDS Alfa Aesar™
3570209
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA3570218 SDS Alfa Aesar™
3570218
50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA3570230 SDS Alfa Aesar™
3570230
250g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

  • Ampouled under argon
Description & Specifications

Chemical Identifiers

10099-58-8
MFCD00011068
14,5363
245.27
UN3077
Lanthanum(III) chloride, Ultra dry

Specifications

245.27
Ampule
Beads
10g
Hygroscopic
MFCD00011068
860°C
−10 Mesh
99.9%, (REO)
1,000°C
UN3077
3.842g/mL
Odorless
White
14,5363
10099-58-8
Lanthanum(III) chloride, Ultra dry

Safety and Handling

GHS H Statement
H318-H290-H317
Causes serious eye damage.
May be corrosive to metals.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P305+P351+P338-P310-P406-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : OE4375000

TSCA : TSCA

storageNote1 :

  • Ampouled under argon