missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lanthanum(III) chloride heptahydrate, ACS, Alfa Aesar™

Manufacturer:  Alfa Aesar 1291630

 View more versions of this product

Catalog No. AA1291630



Specifications

Specifications

Lanthanum(III) chloride heptahydrate
10025-84-0
MFCD00149756
371.37 (245.27 Anhydrous)
ACS
91°C (decomposition)
For analysis ACS
LaCl3·7H2O
14,5363
Crystalline
Odorless
250g

Chemical Identifiers

10025-84-0
LaCl3·7H2O
MFCD00149756
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : OE4470000

TSCA : TSCA

Documents