missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lanthanum(III) chloride heptahydrate, 99.99% (REO), Alfa Aesar™, REacton™

$96.53 - $1,081.84

Chemical Identifiers

CAS 10025-84-0
Molecular Formula LaCl3·7H2O
MDL Number MFCD00149756
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1126818
View Documents
Thermo Fisher Scientific
1126818
50g Each for $96.53
 
AA1126830
View Documents
Thermo Fisher Scientific
1126830
250g Each for $367.73
 
AA11268A1
View Documents
Thermo Fisher Scientific
11268A1
1kg Each for $1,081.84
 
Specifications

Chemical Identifiers

10025-84-0
LaCl3·7H2O
MFCD00149756

Specifications

10025-84-0
LaCl3·7H2O
White wet crystalline aggregates
371.37 (245.27 Anhydrous)
Odorless
91°C (decomposition)
Lanthanum(III) chloride heptahydrate
99.99% (REO)
MFCD00149756
Soluble in water,alcohol and acids.
Wet Crystalline Aggregates
White
50g
Safety and Handling

Safety and Handling

P234-P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P390-P501c

H290-H317-H318

EINECSNumber : 233-237-5

RTECSNumber : OE4470000

TSCA : Yes

Recommended Storage : Ambient temperatures