Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum(III) chloride, anhydrous, 99.9% (REO)

$42.50 - $152.19

Chemical Identifiers

Chemical Name or Material Lanthanum (III) chloride
CAS 10099-58-8
Name Note Anhydrous
Molecular Formula LaCl3
Formula Weight 245.27
MDL Number MFCD00011068
UN Number UN3077
Merck Index 14,5363
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA8791114

  SDS
alfa aesar™
8791114*
25g Each for $152.19

AA8791106

  SDS
alfa aesar™
8791106*
5g Each for $42.50
Description & Specifications

Specifications

Boiling Point 1,000°C
Formula Weight 245.27
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P310-P406-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
tore in corrosive resistant/... container with a resistant inner liner.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 860°C
Assay Percent Notes (REO)
UN Number UN3077
CAS 10099-58-8
Assay 99.9%
Molecular Formula LaCl3
MDL Number MFCD00011068
TSCA TSCA
Quantity 25g
Health Hazard 1 Danger
Density 3.842g/mL
Name Note Anhydrous
Physical Form Powder
Merck Index 14,5363
Color White
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Chemical Name or Material Lanthanum (III) chloride
Odor Odorless
RTECS Number OE4375000
Health Hazard 2 GHS H Statement
H318-H290-H317
Causes serious eye damage.
May be corrosive to metals.
May cause an allergic skin reaction.
Sensitivity Hygroscopic
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P310-P406-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
tore in corrosive resistant/... container with a resistant inner liner.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H290-H317
Causes serious eye damage.
May be corrosive to metals.
May cause an allergic skin reaction.