Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum(III) carbonate hydrate, REacton™, 99.99% (REO)

Chemical Identifiers

Chemical Name or Material Lanthanum(III) carbonate hydrate
CAS 54451-24-0
Purity Grade Reacton™
Formula Weight 457.85 (Anhydrous)
MDL Number MFCD00011069
Merck Index 14,5364
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1319718

  SDS
alfa aesar™
1319718
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1319730

  SDS
alfa aesar™
1319730
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 457.85 (Anhydrous)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material Lanthanum(III) carbonate hydrate
Health Hazard 1 Warning
Sensitivity Hygroscopic
Density 2.6g/mL to 2.7g/mL
CAS 54451-24-0
Quantity 50g
Assay 99.99%
TSCA TSCA
Merck Index 14,5364
MDL Number MFCD00011069
Assay Percent Notes (REO)
Physical Form Powder/Lump
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Purity Grade Reacton™
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.