missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum(III) carbonate hydrate, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1663914

 View more versions of this product

Catalog No. AA1663914



Specifications

Specifications

Lanthanum(III) carbonate hydrate
99.9%
MFCD00011069
457.85 (Anhydrous)
2.6g/mL to 2.7g/mL
25g
54451-24-0
(REO)
14,5364
Powder
Hygroscopic

Chemical Identifiers

54451-24-0
MFCD00011069
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents