missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum(III) bromide hydrate, REacton™, 99.99% (REO)

Manufacturer:  Alfa Aesar™ 1319830

 View more versions of this product

Catalog No. AA1319830



Specifications

Specifications

Lanthanum(III)bromidehydrate
(REO)
MFCD00798151
Soluble in water.
O.[Br-].[Br-].[Br-].[La+3]
396.632
378.62 (Anhydrous)
99.99%
Hygroscopic
250g
224183-16-8
Br3H2LaO
acmc-1chtf, lanthanum bromide hydrate, lanthanum bromide-water 1/3/1, lanthanum iii bromide hydrate
VVYBKVNBDAPKGO-UHFFFAOYSA-K
lanthanum(3+);tribromide;hydrate
71317322
Crystalline
Reacton™
White

Chemical Identifiers

224183-16-8
396.632
VVYBKVNBDAPKGO-UHFFFAOYSA-K
71317322
O.[Br-].[Br-].[Br-].[La+3]
Br3H2LaO
MFCD00798151
acmc-1chtf, lanthanum bromide hydrate, lanthanum bromide-water 1/3/1, lanthanum iii bromide hydrate
lanthanum(3+);tribromide;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 236-896-7

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents