Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum(III) acetate sesquihydrate, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Lanthanum(III) acetate sesquihydrate
CAS 25721-92-0
Formula Weight 343.05 (316.03 Anhydrous)
MDL Number MFCD00013037
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1529922

 
alfa aesar™
1529922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1529936

 
alfa aesar™
1529936
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 343.05 (316.03 Anhydrous)
Chemical Name or Material Lanthanum(III) acetate sesquihydrate
TSCA TSCA
Physical Form Lump/Powder
Assay 99.9%
Sensitivity Hygroscopic
Assay Percent Notes (REO)
MDL Number MFCD00013037
CAS 25721-92-0
Quantity 100g