Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum(III) 2-methoxyethoxide, 99.9% (REO), 5% w/v in 2-methoxyethanol

Preparation of lanthanum oxide thin films by the Sol-Gel method

Chemical Identifiers

Chemical Name or Material Lanthanum (III) 2-methoxyethoxide
CAS 115685-56-8
Linear Formula La(OCH2CH2OCH3)3
Formula Weight 364.17
MDL Number MFCD00210619
UN Number UN1188
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42522AC

 
alfa aesar™
42522AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 364.17
Density 0.96g/mL
Physical Form Liquid
MDL Number MFCD00210619
UN Number UN1188
Health Hazard 1 Danger
Boiling Point 124°C
Sensitivity Moisture Sensitive
Chemical Name or Material Lanthanum (III) 2-methoxyethoxide
Assay 99.9%
Assay Percent Notes (REO)
Linear Formula La(OCH2CH2OCH3)3
CAS 115685-56-8
Health Hazard 2 GHS H Statement
H360-H226-H302-H312-H332
May damage fertility or the unborn child.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P201-P308+P313
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Color Yellow
Quantity 25mL
Flash Point 40°C (104°F)
Concentration 5% w/v in 2-methoxyethanol
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYLENE GLYCOL MONOMETHYL ETHER
Safety and Handling

Danger

GHS P Statement
P201-P308+P313
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

GHS H Statement
H360-H226-H302-H312-H332
May damage fertility or the unborn child.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.