missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum(III) 2,4-pentanedionate hydrate, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1319422

 View more versions of this product

Catalog No. AA1319422



Specifications

Specifications

Lanthanum(III) 2,4-pentanedionate hydrate
(REO)
MFCD00149057
Insoluble in water.
CC(=CC(=O)C)O.CC(=CC(=O)C)O.CC(=CC(=O)C)O.O.O.[La]
475.286
436.24 (Anhydrous)
99.9%
White to Yellow
100g
64424-12-0
C15H28LaO8
lanthanum acetylacetonate hydrate min
DTGMYUJFAPMLCR-UHFAIHIGSA-N
(Z)-4-hydroxypent-3-en-2-one;lanthanum;dihydrate
132984033
Powder
Hygroscopic
143°C

Chemical Identifiers

64424-12-0
475.286
DTGMYUJFAPMLCR-UHFAIHIGSA-N
132984033
CC(=CC(=O)C)O.CC(=CC(=O)C)O.CC(=CC(=O)C)O.O.O.[La]
C15H28LaO8
MFCD00149057
lanthanum acetylacetonate hydrate min
(Z)-4-hydroxypent-3-en-2-one;lanthanum;dihydrate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 238-187-8

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents