Please login to your online account to display your discounted pricing

Lanthanum ICP Standard, 1mL = 1mg La (1,000ppm La), Ricca Chemical


Quantity 100mL
Packaging Poly natural

Manufacturer: ricca chemical company  RIPLA1KN100

 View more versions of this product

Catalog No. PLA1KN-100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 100mL
Chemical Name or Material Lanthanum ICP Standard
CAS 7732-18-5
CAS Min % 93.0
Concentration or Composition Notes 1,000ppm La
Equivalence 1 mL = 1 mg La
CAS Min % 4.62
CAS Max % 0.11
Packaging Poly natural
CAS 1312-81-8
CAS Min % 0.11
CAS Max % 4.81
CAS 7697-37-2
CAS Max % 97.0

Specifications

Chemical Name or Material Lanthanum ICP Standard
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 7697-37-2
CAS Min % 4.62
CAS Max % 4.81
CAS 1312-81-8
CAS Min % 0.11
CAS Max % 0.11
Physical Form Liquid