missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum foil, 1.0mm (0.04 in.) thick, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 10220FF

 View more versions of this product

Catalog No. AA10220FF



Description

Description

  • Packaged under argon
Specifications

Specifications

Lanthanum foil
25 x 25mm
7439-91-0
La
UN3208
FZLIPJUXYLNCLC-UHFFFAOYSA-N
lanthanum
Mylar under Argon
CHEBI:33336
99.9%
≈3.9g/25x25mm
1.0mm (0.04 in.) thick
(REO)
MFCD00011066
2+ ion
[La]
138.905
23926
Foil
Silver to Gray

Chemical Identifiers

7439-91-0
138.905
FZLIPJUXYLNCLC-UHFFFAOYSA-N
23926
lanthanum
La
MFCD00011066
2+ ion
CHEBI:33336
[La]
Safety and Handling

Safety and Handling

GHS H Statement
H228-H261
Flammable solid.
In contact with water releases flammable gas.

GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.

TSCA : TSCA

storageNote1 :

  • Packaged under argon

Documents