Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum chromite, 99.5% (REO)

Chemical Identifiers

Chemical Name or Material Lanthanum chromite
CAS 12017-94-6
Formula Weight 238.9
MDL Number MFCD00053990
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2278509

  SDS
alfa aesar™
2278509
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA2278518

  SDS
alfa aesar™
2278518
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 238.9
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Lanthanum chromite
TSCA TSCA
Mesh Size −200 Mesh
Odor Odorless
Physical Form Powder
Assay 99.5%
Assay Percent Notes (REO)
MDL Number MFCD00053990
CAS 12017-94-6
Melting Point 2450°C
Quantity 10g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Density 5.6g/mL
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.