Please login to your online account to display your discounted pricing

Lanthanum Chloride Heptahydrate (Lumps/Certified), Fisher Chemical


Quantity 50g
Packaging Poly Bottle

Manufacturer: fisher scientific company  L950

 View more versions of this product

Catalog No. L9-50

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Formula Weight 371.37
Melting Point 91°C
Quantity 50g
Physical Form Solid
Molecular Formula Cl3La · 7 H2O
Viscosity [NULL]
Decomposition Information >91°C
Color White
Chemical Name or Material Lanthanum Chloride Heptahydrate
Identification Pass Test
Magnesium (Mg) 1ppm max.
Packaging Poly Bottle
MDL Number MFCD00149756
CAS 10025-84-0
ChemAlert Storage Symbol Gray
Additional Information Vapor Pressure: Negligible
Calcium (Ca) 0.001% max.

Specifications

Chemical Name or Material Lanthanum Chloride Heptahydrate
CAS 10025-84-0
Purity Grade Certified Grade
Physical Form Solid
Molecular Formula Cl3La · 7 H2O
Formula Weight 371.37
MDL Number MFCD00149756
Accessories
 

SDS & Safety Documents