missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lanthanum Chloride Heptahydrate (Lumps/Certified), Fisher Chemical

Manufacturer:  Thermo Scientific L9250

 View more versions of this product

Catalog No. L9-250




Specifications

Specifications

Lanthanum Chloride Heptahydrate
Vapor Pressure: Negligible
MFCD00149756
Solid
0.001% max.
1ppm max.
>91°C
91°C
10025-84-0
Cl3La · 7 H2O
371.37
Certified
Pass Test
Poly Bottle
White
250g

Chemical Identifiers

10025-84-0
Cl3La · 7 H2O
MFCD00149756
Safety and Handling

Safety and Handling

SDS Documents