Please login to your online account to display your discounted pricing

Lanthanum Chloride Heptahydrate (Lumps/Certified), Fisher Chemical

$247.54 - $856.54

Chemical Identifiers

Chemical Name or Material Lanthanum Chloride Heptahydrate
CAS 10025-84-0
Purity Grade Certified Grade
Physical Form Solid
Molecular Formula Cl3La · 7 H2O
Formula Weight 371.37
MDL Number MFCD00149756
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

L9-50

  SDS
fisher scientific company
L950
Poly Bottle 50g Each for $247.54
Case of 6 Each for $1,485.24

L9-250

  SDS
fisher scientific company
L9250
Poly Bottle 250g Each for $856.54
Case of 6 Each for $5,139.24
Description & Specifications

Specifications

Purity Grade Certified Grade
Melting Point 91°C
Formula Weight 371.37
Quantity 50g
Physical Form Solid
Molecular Formula Cl3La · 7 H2O
Color White
Decomposition Information >91°C
Identification Pass Test
Packaging Poly Bottle
ChemAlert Storage Symbol Gray
Chemical Name or Material Lanthanum Chloride Heptahydrate
CAS 10025-84-0
MDL Number MFCD00149756
Calcium (Ca) 0.001% max.
Additional Information Vapor Pressure: Negligible
Magnesium (Mg) 1ppm max.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling