Please login to your online account to display your discounted pricing

Lanthanum Chloride, 7-Hydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$380.91 - $1,317.31

Chemical Identifiers

Chemical Name or Material Lanthanum Chloride
CAS 10099-58-8
Name Note 7-Hydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula LaCl3·7H2O
Formula Weight 371.38
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003311

 
avantor performance
225505
Wide Mouth Amber Glass Bottle 100g Each for $380.91

02003312

 
avantor performance
225507
Wide Mouth Amber Glass Bottle 500g Each for $1,317.31
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 371.38
Chemical Name or Material Lanthanum Chloride
Quantity 100g
Name Note 7-Hydrate
CAS 10099-58-8
Molecular Formula LaCl3·7H2O
Packaging Wide Mouth Amber Glass Bottle

    For Flame Enhancement in Atomic Absorption Spectroscopy