Please login to your online account to display your discounted pricing

Lanthanum Chloride, 7-Hydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Packaging Wide Mouth Amber Glass Bottle

Manufacturer: avantor performance  225507

 View more versions of this product

Catalog No. 02003312

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 371.38
Chemical Name or Material Lanthanum Chloride
Molecular Formula LaCl3·7H2O
CAS 10099-58-8
Quantity 500g
Name Note 7-Hydrate
Packaging Wide Mouth Amber Glass Bottle

Specifications

Chemical Name or Material Lanthanum Chloride
CAS 10099-58-8
Name Note 7-Hydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula LaCl3·7H2O
Formula Weight 371.38

    For Flame Enhancement in Atomic Absorption Spectroscopy