Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum boride, 99.5% (REO)

Chemical Identifiers

Chemical Name or Material Lanthanum boride
CAS 12008-21-8
Formula Weight 203.78
MDL Number MFCD00151350
Synonym Lanthanum hexaboride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4032509

  SDS
alfa aesar™
4032509
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4032518

  SDS
alfa aesar™
4032518
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4032530

  SDS
alfa aesar™
4032530
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 203.78
Chemical Name or Material Lanthanum boride
TSCA TSCA
Mesh Size −325 Mesh
Odor Odorless
Physical Form Powder
Assay 99.5%
Assay Percent Notes (REO)
MDL Number MFCD00151350
CAS 12008-21-8
Synonym Lanthanum hexaboride
Melting Point 2210°C
Quantity 10g
Solubility Information Insoluble in water, HCl
Density 2.61g/mL