Please login to your online account to display your discounted pricing

Lanthanum Aluminum Oxide Substrate, 10 x 10 x 1mm, Polished One Side, 100 Orientation, Alfa Aesar™

100 Orientation

$210.88 - $955.89

Chemical Identifiers

Chemical Name or Material Lanthanum Aluminum Oxide Substrate
Name Note Polished Two Sides
CAS 12003-65-5
Physical Form Plate
Molecular Formula LaAlO3
MDL Number MFCD00144550
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38495KT

 
alfa aesar
38495KT
1 ea. Each for $210.88

AA38495KU

 
alfa aesar
38495KU
5 ea. Each for $955.89
Description & Specifications

Specifications

Physical Form Plate
Quantity 1 ea.
Color Yellow-brown
Chemical Name or Material Lanthanum Aluminum Oxide Substrate
Odor Odorless
Name Note Polished Two Sides
Molecular Formula LaAlO3
MDL Number MFCD00144550
CAS 12003-65-5