missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lanthanum Aluminum Oxide Substrate, 10 x 10 x 0.5mm, Polished Two Sides, 100 Orientation, Alfa Aesar™

100 Orientation

$157.00 - $680.36

Chemical Identifiers

CAS 12003-65-5
Physical Form Plate
Molecular Formula LaAlO3
MDL Number MFCD00144550
Chemical Name or Material Lanthanum Aluminum Oxide Substrate, Polished Two Sides
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA41434KT SDS Alfa Aesar
41434KT
1 ea. Each for $157.00
AA41434KU SDS Alfa Aesar
41434KU
5 ea. Each for $680.36
Specifications Description & Specifications

Chemical Identifiers

12003-65-5
LaAlO3
Lanthanum Aluminum Oxide Substrate, Polished Two Sides
Plate
MFCD00144550

Specifications

Plate
Yellow-brown
LaAlO3
Odorless
1 ea.
12003-65-5
MFCD00144550
Lanthanum Aluminum Oxide Substrate, Polished Two Sides