Please login to your online account to display your discounted pricing

Alfa Aesar™ Lanthanum aluminum oxide, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Lanthanum aluminum oxide
CAS 12003-65-5
Formula Weight 213.88
MDL Number MFCD00144550
Synonym Lanthanum aluminate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3925006

  SDS
alfa aesar™
3925006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3925014

  SDS
alfa aesar™
3925014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 213.88
Odor Odorless
Assay 99.9%
Quantity 5g
Chemical Name or Material Lanthanum aluminum oxide
Physical Form Powder
MDL Number MFCD00144550
CAS 12003-65-5
Synonym Lanthanum aluminate
Assay Percent Notes (metals basis)