missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Landiolol Hydrochloride, TCI America™

Manufacturer:  TCI America L0296250MG

 View more versions of this product

Catalog No. L0296250MG



Specifications

Specifications

Landiolol Hydrochloride
126°C
144481-98-1
MFCD00911043
[H+].CC1(OCC(O1)COC(=O)CCC2=CC=C(C=C2)OCC(CNCCNC(=O)N3CCOCC3)O)C.[Cl-]
546.058
546.06
≥98.0% (HPLC,T)
White
250mg
C25H40ClN3O8
DLPGJHSONYLBKP-IKGOIYPNSA-N
[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methyl 3-[4-[(2S)-2-hydroxy-3-[2-(morpholine-4-carbonylamino)ethylamino]propoxy]phenyl]propanoate;hydron;chloride
46856312
Crystal-Powder at 20°C

Chemical Identifiers

144481-98-1
546.058
DLPGJHSONYLBKP-IKGOIYPNSA-N
[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methyl 3-[4-[(2S)-2-hydroxy-3-[2-(morpholine-4-carbonylamino)ethylamino]propoxy]phenyl]propanoate;hydron;chloride
C25H40ClN3O8
MFCD00911043
46856312
[H+].CC1(OCC(O1)COC(=O)CCC2=CC=C(C=C2)OCC(CNCCNC(=O)N3CCOCC3)O)C.[Cl-]
Safety and Handling

Safety and Handling

RTECSNumber : DA8346700

TSCA : No

Documents