Please login to your online account to display your discounted pricing

American Educational Products Landforms Model

A river runs through it — as well as mountains, deserts and canyons.

$14.06 - $867.71

Specifications

For Use With (Application) Gives geologic and geo graphic details
Grade 5 to 12
View More Specs
Includes:  Eight models and seven coordinated audio tapes
Products
Catalog Number Mfr. No. Product Type Price Quantity    

S46995-1

 
american educational products
HUB790
Landform discovery pack Each for $867.71

S46995-1D

 
american educational products
HUB511
Student workbook Each for $14.06
Description & Specifications

Specifications

For Use With (Application) Gives geologic and geo graphic details
Grade 5 to 12

  • Natural-color, durable, raised-relief models display more than 100 key land-forming features
  • Used separately, models isolate the features on glaciers, coastlines, canyons, deserts, lakes and mountains
  • Models join for a unique visual comparison, 39 in. (1m) across
  • Lesson plan provides exercises, supplemental activities and a glossary that is keyed to features found on the landform models