Please login to your online account to display your discounted pricing

LAMP5 Rat anti-Human, Clone: 34.2, eBioscience™

Rat Monoclonal Antibody

$175.45 - $175.45

Specifications

Antigen LAMP5
Clone 34.2
Host Species Rat
Species Reactivity Human
Applications ELISA
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122865

 
life technologies
LS14977880
25μg Each for $175.45
Description & Specifications

Specifications

Antigen LAMP5
Clone 34.2
Host Species Rat
Species Reactivity Human
Applications ELISA
Applications Flow Cytometry (Intracellular Staining)
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody 34.2 recognizes human lysosome-associated membrane protein 5 (LAMP5), a member of the LAMP family. LAMP5 differs from other LAMP family members in that its expression is more restricted and is referred to as brain and dendritic cell-associated LAMP-like molecule (BAD-LAMP). In humans, BAD-LAMP is expressed in post-natal neurons and non-activated plasmacytoid dendritic cells. Expression in neurons is localized to intracellular vesicles distributed in specific microdomains along neurites and may play a role in endocytosis. BAD-LAMP is localized to the endoplasmic reticulum-Golgi intermediate compartment (ERGIC) of plasmacytoid dendritic cells and is lost upon TLR activation.