Please login to your online account to display your discounted pricing

LAMP5 Rat anti-Human, Alexa Fluor 488, Clone: 34.2, eBioscience™

Rat Monoclonal Antibody

$175.45 - $442.25

Specifications

Antigen LAMP5
Clone 34.2
Host Species Rat
Species Reactivity Human
Applications Immunohistochemistry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124647

 
life technologies
LS53977880
25μg Each for $175.45

501124648

 
life technologies
LS53977882
100μg Each for $442.25
Description & Specifications

Specifications

Antigen LAMP5
Clone 34.2
Host Species Rat
Species Reactivity Human
Applications Immunohistochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Regulatory Status RUO
Conjugate Alexa Fluor 488
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody 34.2 recognizes human lysosome-associated membrane protein 5 (LAMP5), a member of the LAMP family. LAMP5 differs from other LAMP family members in that its expression is more restricted and is referred to as brain and dendritic cell-associated LAMP-like molecule (BAD-LAMP). In humans, BAD-LAMP is expressed in post-natal neurons and non-activated plasmacytoid dendritic cells. Expression in neurons is localized to intracellular vesicles distributed in specific microdomains along neurites and may play a role in endocytosis. BAD-LAMP is localized to the endoplasmic reticulum-Golgi intermediate compartment (ERGIC) of plasmacytoid dendritic cells and is lost upon TLR activation.