missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

LAMP5 Rat anti-Human, Alexa Fluor 488, Clone: 34.2, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS53977882

 View more versions of this product

Catalog No. 501124648



Description

Description

The monoclonal antibody 34.2 recognizes human lysosome-associated membrane protein 5 (LAMP5), a member of the LAMP family. LAMP5 differs from other LAMP family members in that its expression is more restricted and is referred to as brain and dendritic cell-associated LAMP-like molecule (BAD-LAMP). In humans, BAD-LAMP is expressed in post-natal neurons and non-activated plasmacytoid dendritic cells. Expression in neurons is localized to intracellular vesicles distributed in specific microdomains along neurites and may play a role in endocytosis. BAD-LAMP is localized to the endoplasmic reticulum-Golgi intermediate compartment (ERGIC) of plasmacytoid dendritic cells and is lost upon TLR activation.
Specifications

Specifications

LAMP5
34.2
Alexa Fluor 488
Conjugated
IgG1, kappa
RUO
Primary
Human
Immunohistochemistry, Immunohistochemistry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Rat
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Documents

For Research Use Only.