Please login to your online account to display your discounted pricing

LaMotte™ Turbidity Standards


For use with LaMotte Portable Turbidity Meters

Manufacturer: lamotte™  1455

 View more versions of this product

Catalog No. 03-920-558

  • / Each
 

Description & Specifications

Specifications

Turbidity 100 FNU Standard (ISO), 60mL
Volume (English) 2.02 oz.
Volume (Metric) 60mL
Type Turbidity Standard
Certifications/Compliance ISO
For Use With (Equipment) For 2020WI portable turbidity meter