LaMotte™ SMART™ Spectro Reagent Systems
A wide range of reagent systems to meet your quality testing needs
$85.11 - $184.51
Specifications
For Use With SMART™ spectro spectrophotometer
Type LaMotte test reagent
View More Specs
Requires: Use with the Lamotte SMART 3 Colorimeter and the SMART Spectro Spectrophotometer.
Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Range Reagents Test Method Price Quantity  
Catalog Number Mfr. No. Packaging Range Reagents Test Method Price Quantity  
S95115 LaMotte™
3667SC/EMD
50 0 to 3 ppm 6 Zincon Each for $171.45
22-278-518 LaMotte™
3658-SC
50 0 to 0.7 ppm 3 PAN Each for $184.51
22-278-584 LaMotte™
364101SC
50 0 to 0.3 ppm 4 Eriochrome Cyanine R Each for $137.07
22-278-585 LaMotte™
365901SC
25 0 to 1ppm 3 Salicylate Each for $85.11
Specifications
Description
Choose from the following test factors:
  • Zinc Laboratory Reagent (LR)
  • Manganese
  • Aluminum
  • Ozone
  • Cyanide
  • Phosphate
  • Dissolved Oxygent
  • Chromium Hexavalent
  • Ammonia Nitrogen LR
Description & Specifications
Specifications
SMART™ spectro spectrophotometer
LaMotte test reagent