LaMotte™ SMART Spectro™ Spectrophometer and SMART 3™ Colorimeter Reagent Systems
For water analysis in the field or in the lab
Supplier: LaMotte™ 3659CN02SC
Description
Choose from the following test factors:
- Zinc Laboratory Reagent (LR)
- Manganese
- Aluminum
- Ozone
- Cyanide
- Phosphate
- Dissolved Oxygent
- Chromium Hexavalent
- Ammonia Nitrogen LR
Order Info
Additional reagent systems are available. Contact your Fisher Scientific Customer Service Representative for details.
Specifications
Ammonia Nitrogen-LR (Fresh water) | |
SMART Spectro Spectrophometer and SMART 3 Colorimeter | |
0 to 1ppm | |
Ammonia Nitrogen, Low Range (salt water) |
Water testing and environmental studies | |
50 | |
Salicylate |