Please login to your online account to display your discounted pricing

LaMotte™ Reagent Refills


Testing supplies for the discerning scientist

Manufacturer: lamotte™  4498L

 View more versions of this product

Catalog No. 03-920-351

  • / Each
 

Description & Specifications

Specifications

Shelf Life 18 Months
Volume (Metric) 500mL
Storage Requirements 25°C
Volume (English) 16.9 oz.
Product Type Chlorine Reagent 1
Accessories

18 Months