Please login to your online account to display your discounted pricing

LaMotte™ Reagent Refills


Testing supplies for the discerning scientist

Manufacturer: lamotte™  4636H

 View more versions of this product

Catalog No. 03-920-461

  • / Each
 

Description & Specifications

Specifications

Storage Requirements 25°C
Volume (English) 2.02 oz.
Volume (Metric) 60mL
Shelf Life 3 Years
Product Type Sulfide Test 4