LaMotte™ LTC3000w Series Laboratory Turbidity, Chlorine and Color Meter
Bench-top turbidimeter for turbidity, chlorine and color measurement
Manufacturer:  LaMotte™ 1972ISO
 View more versions of this product
Catalog No. 03-920-562
Specifications Description & Specifications
Specifications
0.01 NTU/FNU, 0.1 NTU/FNU, 1 NTU/FNU
0,1 and 10 NTU standards, sample bottle, 6 sample tubes, 100 each free and total chlorine DPD tablets and USB cable and computer/wall adapter
LED
8.75 in.
3 in.
22.2cm
0.68kg
3.7V
ISO
Laboratory Turbidity And Chlorine Meter
<2 sec.
1.5 lb.
For turbidity, chlorine and color measurement
860mm LED
7.6cm
Lithium ion rechargeable