Please login to your online account to display your discounted pricing

LaMotte™ Insta-Test™ Single and Multi-Factor Test Strips


For water quality testing

Manufacturer: lamotte™  299812

 View more versions of this product

Catalog No. 03-920-404

  • / Dozen of 12 Each
 

Description & Specifications

Specifications

For Use With (Application) Water quality testing
Storage Requirements Store tightly sealed vial in a cool, dry storage area away from moisture; Keep out of direct sunlight
Quantity 10 Papers/blister card; 10 Cards/pack
Parameter Specifications Sodium Chloride
Range 1500 to 5000ppm
Product Type Single Factor Test Strips