Please login to your online account to display your discounted pricing

LaMotte™ Insta-Test™ Single and Multi-Factor Test Strips


For water quality testing

Manufacturer: lamotte™  2964G

 View more versions of this product

Catalog No. 03-920-400

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Water quality testing
Parameter Specifications Chlorine - free, low range
Quantity 25 Strips per Vial
Range 0-0.5-1-3-5-10ppm
Storage Requirements Store tightly sealed vial in a cool, dry storage area away from moisture; Keep out of direct sunlight
Product Type Single Factor Test Strips