Please login to your online account to display your discounted pricing

LaMotte™ Insta-Test™ Single and Multi-Factor Test Strips


For water quality testing

Manufacturer: lamotte™  2987RSD

 View more versions of this product

Catalog No. 03-920-426

  • / Pack
 

Description & Specifications

Specifications

For Use With (Application) Water quality testing
Storage Requirements Store tightly sealed vial in a cool, dry storage area away from moisture; Keep out of direct sunlight
Parameter Specifications Total Chlorine and pH
Product Type Multi-Factor Test Strips
Range Chlorine 0-1-5-10-20-50ppm, pH 4-5-6-7-8-9-10
Quantity 25 Tests/vial; 12 Vials/case